Learn More
Description
Specifications
Specifications
| Antigen | LC3A |
| Applications | Chemotaxis, ChIP Assay, ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount), Immunoprecipitation, Simple Western, Southern Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2.0 ug/ml, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature (PMID 33035707), Flow Cytometry reported in scientific literature (PMID 24419333), ELISA reported in scientific literature (PMID 20930550), Immunohistochemistry 1:200-1:400, Immunocytochemistry/Immunofluorescence 1:100-1:300. Use reported in scientific literature (PMID 21545732), Immunoprecipitation 20 ug/ 500 ug of lysate, Immunohistochemistry-Paraffin 1:200-1:400. Use reported in scientific literature (PMID 26571030), Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 28253371), Southern Blot, Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 31783118), Chromatin Immunoprecipitation (ChIP) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | Q9GZQ8 |
| Gene Alias | ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A |
| Gene Symbols | MAP1LC3A |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
