Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3A Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002331SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002331SS 0.025 mL
NB1002331 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002331SS Supplier Novus Biologicals Supplier No. NB1002331SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 265 publications

LC3A Polyclonal specifically detects LC3A in Amphibian, Bovine, Canine, Fish, Human, Mouse, Plant, Rat, SARS-CoV, Zebrafish samples. It is validated for Chemotaxis, ChIP Assay, ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount, Immunoprecipitation, Simple Western, Southern Blot, Western Blot.

Specifications

Antigen LC3A
Applications Chemotaxis, ChIP Assay, ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount), Immunoprecipitation, Simple Western, Southern Blot, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2.0 ug/ml, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature (PMID 33035707), Flow Cytometry reported in scientific literature (PMID 24419333), ELISA reported in scientific literature (PMID 20930550), Immunohistochemistry 1:200-1:400, Immunocytochemistry/Immunofluorescence 1:100-1:300. Use reported in scientific literature (PMID 21545732), Immunoprecipitation 20 ug/ 500 ug of lysate, Immunohistochemistry-Paraffin 1:200-1:400. Use reported in scientific literature (PMID 26571030), Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 28253371), Southern Blot, Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 31783118), Chromatin Immunoprecipitation (ChIP)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9GZQ8
Gene Alias ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A
Gene Symbols MAP1LC3A
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human LC3 protein sequence (between residues 25-121). [Uniprot: Q9H492].
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, Cytoskeleton Markers, Hypoxia, Macroautophagy, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84557
Test Specificity This antibody detects both LC3A and LC3B.
Target Species Amphibian, Mouse, Bovine, Canine, Fish, Fish, Human, Zebrafish, Mouse, Plant, Rat, SARS-CoV, Zebrafish
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.