Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3A Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB1002331R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002331R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002331R Supplier Novus Biologicals Supplier No. NB1002331R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LC3A Polyclonal specifically detects LC3A in Amphibian, Canine, Fish, Human, Mouse, Plant, Rat, Zebrafish samples. It is validated for ChIP Assay, ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount, Immunoprecipitation, Southern Blot, Western Blot.

Specifications

Antigen LC3A
Applications ChIP Assay, ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount), Immunoprecipitation, Southern Blot, Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Whole-Mount
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9GZQ8
Gene Alias ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A
Gene Symbols MAP1LC3A
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human LC3 protein sequence (between residues 25-121). [Uniprot: Q9H492].
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, Cytoskeleton Markers, Hypoxia, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84557
Test Specificity This antibody detects both LC3A and LC3B.
Target Species Amphibian, Canine, Fish, Human, Mouse, Plant, Rat, Zebrafish
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.