Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3A Antibody, Novus Biologicals™
SDP

Catalog No. NBP224394 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224394 0.1 mg
NBP224394SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224394 Supplier Novus Biologicals Supplier No. NBP224394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LC3A Polyclonal specifically detects LC3A in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen LC3A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000. Use reported by customer review, Immunohistochemistry, Immunohistochemistry-Paraffin 5 - 10 ug/mL
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9H492
Gene Alias ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A
Gene Symbols MAP1LC3A
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human ATG8 (LC3, MAP1LC3A, APG8) was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, Cytoskeleton Markers, Hypoxia, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84557
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, dog and mouse, 93% homologous in cow and Xenopus, 80% homologous in opossum and 73% homologous in pufferfish.
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.