Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3B Antibody (1251B), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB001454 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001454 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001454 Supplier Novus Biologicals Supplier No. NBP260735APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LC3B Monoclonal specifically detects LC3B in Human, Mouse, Rat samples. It is validated for Flow (Intracellular), Knockout Validated.

Specifications

Antigen LC3B
Applications Flow Cytometry
Classification Monoclonal
Clone 1251B
Conjugate APC
Dilution Flow (Intracellular)
Formulation PBS with 0.05% Sodium Azide
Gene Alias ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B
Gene Symbols MAP1LC3B
Host Species Rabbit
Immunogen Recombinant monoclonal LC3B Antibody (1251B) was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [UniProt# Q9GZQ8].
Molecular Weight of Antigen 14.688 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Mitophagy, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 81631
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.