Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3B Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB6001384V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001384V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001384V Supplier Novus Biologicals Supplier No. NB6001384V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LC3B Polyclonal specifically detects LC3B in Human, Mouse, Rat, Porcine, Bacteria, Bovine, Canine, Primate, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Free-Floating, Knockout Validated, Knockdown Validated.

Specifications

Antigen LC3B
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunohistochemistry (Free Floating), Immunoassay, KnockDown
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Free-Floating, Knockout Validated, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9GZQ8
Gene Alias ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B
Gene Symbols MAP1LC3B
Host Species Rabbit
Immunogen Polyclonal LC3B Antibody was made to a synthetic peptide made to the N-terminal region of the human LC3B protein. [Uniprot: Q9GZQ8]
Molecular Weight of Antigen 14.688 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Mitophagy, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 81631
Test Specificity Human, mouse, rat and bovine. Zebrafish reactivity reported in scientific literature (PMID: 23724125). Canine and primate reactivity reported in scientific literature (PMID: 24027311) Porcine reactivity reported in scientific literature (PMID: 25378587) Other species have not been tested.
Target Species Human, Mouse, Rat, Pig, Bacteria, Bovine, Canine, Primate, Zebrafish
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.