Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3C Antibody, Novus Biologicals™
SDP

Catalog No. NB393878 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB393878 25 μL
NBP257604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB393878 Supplier Novus Biologicals Supplier No. NBP25760425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LC3C Polyclonal specifically detects LC3C in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen LC3C
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias ATG8J, Autophagy-related protein LC3 C, Autophagy-related ubiquitin-like modifier LC3 C, LC3C, LC3-like protein 2, MAP1 light chain 3-like protein 2, MAP1 light chain 3-like protein 3, MAP1A/MAP1B LC3 C, microtubule-associated protein 1 light chain 3 gammaMAP1A/MAP1B light chain 3 C, microtubule-associated proteins 1A/1B light chain 3C
Gene Symbols MAP1LC3C
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TKFLVPQELTMTQFLSIIRSRMVLRATEAF
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Autophagy, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 440738
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.