Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LCA5L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP170597
Description
LCA5L Polyclonal specifically detects LCA5L in Human, Mouse samples. It is validated for Western Blot.Specifications
LCA5L | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
LCA5L | |
Synthetic peptides corresponding to C21ORF13 The peptide sequence was selected from the N terminal of C21ORF13. Peptide sequence SLADLTKTNIDEHFFGVALENNRRSAACKRSPGTGDFSRNSNASNKSVDY. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Olive baboon: 100%; Crab-eating macaque: 100%; Human: 100%;. | |
Human, Mouse, Yeast | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
C21orf13, Leber congenital amaurosis 5-like, leber congenital amaurosis 5-like protein, Lebercilin-like protein, MGC33295 | |
Rabbit | |
Affinity purified | |
RUO | |
150082 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction