Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCAT Antibody, Novus Biologicals™
SDP

Catalog No. NB400107SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400107SS 0.025 mL
NB400107 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400107SS Supplier Novus Biologicals Supplier No. NB400107SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

LCAT Polyclonal specifically detects LCAT in Human, Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen LCAT
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:500
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. P04180
Gene Alias EC 2.3.1.43, lecithin-cholesterol acyltransferasePhospholipid-cholesterol acyltransferase, phosphatidylcholine-sterol acyltransferase
Gene Symbols LCAT
Host Species Rabbit
Immunogen Carboxyl-terminal histidine-tagged human recombinant LCAT purified from the medium of CHO cells overexpressing human LCAT cDNA. [UniProt# P04180]
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3931
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.