Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCAT Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB353674 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB353674 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB353674 Supplier Novus Biologicals Supplier No. NBP300108PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LCAT Polyclonal antibody specifically detects LCAT in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen LCAT
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 2.3.1.43, lecithin-cholesterol acyltransferasePhospholipid-cholesterol acyltransferase, phosphatidylcholine-sterol acyltransferase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human LCAT (Accession#: NP_000220.1; Met1-Glu440)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3931
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.