Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lck Antibody (693010), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB371632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371632 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371632 Supplier Novus Biologicals Supplier No. FAB37041L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lck Monoclonal specifically detects Lck in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Western Blot.

Specifications

Antigen Lck
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 693010
Conjugate DyLight 550
Dilution Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10, EC 2.7.10.2, Leukocyte C-terminal Src kinase, LSK, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16
Host Species Mouse
Immunogen E. coli-derived recombinant human Lck, Met1-Val66, Accession # P06239
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3932
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.