Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCMT1 Antibody (OTI2C9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP271927A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271927A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271927A30 Supplier Novus Biologicals Supplier No. NBP271927AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LCMT1 Monoclonal specifically detects LCMT1 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen LCMT1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2C9
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CGI-68, EC 2.1.1.-, LCMT, leucine carboxyl methyltransferase 1, PPMT1, protein phosphatase methyltransferase 1, Protein-leucine O-methyltransferase
Gene Symbols LCMT1
Host Species Mouse
Immunogen Full length recombinant protein of human LCMT1 (NP_057393) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51451.0
Target Species Human, Mouse, Rat, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.