Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LCN8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145409
Description
May play a role in male fertility. May act as a retinoid carrier protein within the epididymis.
Specifications
LCN8 | |
Polyclonal | |
Unconjugated | |
LCN8 | |
9230106L18Rik; EP17; epididymal 17 kDa lipocalin; Epididymal-specific lipocalin-8; LCN5; LCN8; lipocalin 5; lipocalin 8; mEP17 | |
Rabbit | |
Protein A | |
RUO | |
78076 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
Q924P3 | |
LCN8 | |
Recombinant Mouse LCN8 protein, Leu27-Val176 (Accession #Q924P3). | |
200 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction