Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LCN8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $414.50
Specifications
Antigen | LCN8 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145409
|
Invitrogen™
PA5145409 |
200 μL |
Each for $414.50
|
|
|||||
FEPA5145410
|
Invitrogen™
PA5145410 |
20 μL |
Each for $191.80
|
|
|||||
Description
May play a role in male fertility. May act as a retinoid carrier protein within the epididymis.Specifications
LCN8 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse | |
Q924P3 | |
78076 | |
Recombinant Mouse LCN8 protein, Leu27-Val176 (Accession #Q924P3). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
9230106L18Rik; EP17; epididymal 17 kDa lipocalin; Epididymal-specific lipocalin-8; LCN5; LCN8; lipocalin 5; lipocalin 8; mEP17 | |
LCN8 | |
IgG | |
Protein A | |
LCN8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title