Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCoR Antibody (OTI2F8), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP272299G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272299G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272299G Supplier Novus Biologicals Supplier No. NBP272299G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LCoR Monoclonal specifically detects LCoR in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen LCoR
Applications Western Blot
Classification Monoclonal
Clone OTI2F8
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ38026, KIAA1795RP11-175O19.1, LCoR, ligand dependent nuclear receptor corepressor, Mblk1-related protein 2, MLR2ligand-dependent corepressor
Gene Symbols LCOR
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 204-433 of human LCoR (NP_115816) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84458.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.