Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDLR Antibody (032), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB074540 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB074540 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB074540 Supplier Novus Biologicals Supplier No. NBP290523APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LDLR Monoclonal specifically detects LDLR in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen LDLR
Applications Flow Cytometry
Classification Monoclonal
Clone 32
Conjugate APC
Formulation PBS
Gene Alias FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LDLR (Met 1-Arg 790; NP_034830.2).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3949
Test Specificity No cross-reactivity in ELISA with: Human LDLR/ LDL Receptor
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.