Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDLR Antibody (11A2), FITC, Novus Biologicals™
SDP

Catalog No. NBP222035F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222035F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222035F Supplier Novus Biologicals Supplier No. NBP222035F

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

LDLR Monoclonal specifically detects LDLR in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen LDLR
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 11A2
Conjugate FITC
Dilution Flow Cytometry, ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Gene Symbols LDLR
Host Species Mouse
Immunogen Human recombinant LDLR
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3949
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.