Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDLR Antibody (263123), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB365924 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365924 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365924 Supplier Novus Biologicals Supplier No. FAB2255X
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LDLR Monoclonal specifically detects LDLR in Mouse samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen LDLR
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 263123
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse LDL R, Ala22-Arg790 (Ala23Val, Cys27Gly), Accession # Q6GTJ9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3949
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.