Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ LDN 193188

Click to view available options
Quantity:
10 mg
50 mg
Description
LDN 193188 is an inhibitor of phosphatidylcholine transfer protein (PC-TP). Regulates hepatic glucose metabolism. Also promotes insulin-independent phosphorylation of key insulin signaling molecules.
Specifications
Specifications
| Chemical Name or Material | 2,4-Dichloro-N-[[[4-[[(4,6-dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl]amino]carbonyl]benzamide |
| CAS | 1267610-30-9 |
| Quantity | 50 mg |
| Target | Lipid Metabolism Inhibitors |
| Molecular Formula | C20H17Cl2N5O4S |
| Formula Weight | 494.35 |
| Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction