Learn More
LDS-sample buffer (4X), reducing, Thermo Scientific Chemicals
Description
LDS-sample buffer (4X), reducing is used to prepare protein samples for denaturing gel electrophoresis. It is used in 1D gel electrophoresis, bis-tris gel electrophoresis, tris-acetate gel electrophoresis, protein Gel electrophoresis, proteins, expression, isolation & analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
LDS-sample buffer (4X), reducing is used to prepare protein samples for denaturing gel electrophoresis. It is used in 1D gel electrophoresis, bis-tris gel electrophoresis, tris-acetate gel electrophoresis, protein Gel electrophoresis, proteins, expression, isolation & analysis.
Solubility
Not miscible in water.
Notes
This buffer contains 988mM Tris-HCl, 2.04mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue, and 0.7mM phenol red, and 8% mercaptoethanol at pH 7.5.

Specifications
Specifications
Color | Blue |
Concentration | 4X |
Quantity | 50 mL |
Content And Storage | Ambient Temperatures |
Form | Viscous Liquid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.