Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LE 135
GREENER_CHOICE

Catalog No. 202150
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective RARβ antagonist

Retinoic acid antagonist; displays moderate selectivity for RARβ over RARα (Ki values are 0.22 and 1.4 μM respectively). Highly selective over RARγ and RXRα. Inhibits human HL-60 leukemia cell differentiation induced by Am80 (IC50 = 150 nM).

Chemical Identifiers

CAS 155877-83-1
Molecular Formula C29H30N2O2

Specifications

Quantity 50 mg
Formula Weight 438.56
Purity >97%
Molecular Weight (g/mol) 438.571
Chemical Name or Material LE 135
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.