Alfa Aesar™ Lead bar, approximately 1.35x2.54x30.5cm (0.53x1.0x12.0 in.), 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 00930KJ

Quantity 5 pieces

 View more versions of this product

Catalog No. AA00930KJ

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-92-1
    MDL Number MFCD00134050
    Chemical Name or Material Lead bar, Approx.1.35x2.54x30.5cm (0.53x1.0x12.0 in.)

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    TSCA TSCA
    Physical Form Bar
    CAS 7439-92-1
    Odor Odorless
    Health Hazard 1 Danger
    Quantity 5 pieces
    MDL Number MFCD00134050
    Color Gray
    RTECS Number OF7525000
    Weight ≈1300g/bar
    Chemical Name or Material Lead bar, Approx.1.35x2.54x30.5cm (0.53x1.0x12.0 in.)
    Assay 99.9%, (metals basis)

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit