Learn More
Lead(II) acetate trihydrate, 99%
Description
Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.
Solubility
Freely soluble in water, alcohol and glycerol.
Notes
Decomposes above 200°Air and light sensitive. Incompatible with strong acids and strong oxidizing agents.

Specifications
Specifications
Melting Point | 75°C |
Density | 2.55 |
Quantity | 1000 g |
UN Number | UN1616 |
Beilstein | 3730298 |
Merck Index | 14,5397 |
Solubility Information | Freely soluble in water,alcohol and glycerol. |
Formula Weight | 379.33 (325.28 Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | Lead (II) acetate trihydrate |
Safety and Handling
Hazard Category | H360D-H361f-H372 |
Hazard Statement | GHS H Statement H360-H373 May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE |
EINECSNumber | 206-104-4 |
RTECSNumber | OF8050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.