Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) carbonate, basic, ACS, Thermo Scientific Chemicals

Catalog No. p-7050019
Click to view available options
Quantity:
250 g

Catalog No. AA3332830

Supplier: Thermo Scientific Chemicals 03332830

Only null left
Add to Cart

Ceramic glaze, exterior paint

Lead(II) carbonate is used in ceramic glaze and exterior paint.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1319-46-6
MDL Number MFCD00078155
Synonym White lead; C.I. 77597

Specifications

Melting Point 400°C (decomposition)
Color White
Physical Form Powder
Quantity 250 g
Linear Formula (PbCO3)2·Pb(OH)2
UN Number UN3077
Merck Index 14,1005
Solubility Information Soluble in acids, insoluble in water or alcohol
Formula Weight 775.63
Grade ACS
Odor Odorless
Density 6.14 g/mL
Chemical Name or Material Lead(II) carbonate, Basic
Show More Show Less
Hazard Category H302+H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-290-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.