Learn More
Lead(II) chloride, Puratronic™, 99.999% (metals basis)
Description
Lead chloride is used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells. Analytical reagent, preparation of lead salts, as solder and flux. Pattinson's white lead, pigment in white paint, is the basic lead chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 951°C |
Melting Point | 501°C |
Color | White |
Physical Form | Crystalline |
Quantity | 100 g |
Linear Formula | PbCl2 |
UN Number | UN2291 |
Merck Index | 14,5404 |
Solubility Information | Soluble in hot water,alkali hydroxides and ammonium chloride solution. Insoluble in cold water and alcohol. |
Formula Weight | 278.1 |
Show More |
Safety and Handling
Hazard Category | H302+H332-H350-H360Df-H372 |
Hazard Statement | GHS H Statement H360-H373-H302-H332 May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S. |
EINECSNumber | 231-845-5 |
RTECSNumber | OF9450000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.