Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) chromate, 98%, Thermo Scientific Chemicals

Catalog No. AAA10678A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA10678A1 1 kg
AAA1067830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA10678A1 Supplier Thermo Scientific Chemicals Supplier No. A10678A1
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-97-6
Molecular Formula CrO4Pb
Molecular Weight (g/mol) 323.192
MDL Number MFCD00011158
InChI Key MOUPNEIJQCETIW-UHFFFAOYSA-N
Synonym lead chromate, lead ii chromate, phoenicochroite, plumbous chromate, chrome yellow, chromate de plomb, lead chromate vi, lead chromate pbcro4, unii-aa3229aous
PubChem CID 24460
ChEBI CHEBI:86257
IUPAC Name dioxido(dioxo)chromium;lead(2+)
SMILES [O-][Cr](=O)(=O)[O-].[Pb+2]

Specifications

Melting Point 844°C
Quantity 1 kg
Linear Formula PbCrO4
UN Number UN3077
Merck Index 14,5405
Formula Weight 323.19
Percent Purity 98%
Odor Odorless
Density 6.3
Chemical Name or Material Lead (II) chromate
Hazard Category H350-H360Df-H373
Hazard Statement GHS H Statement
H350-H360-H373-H201
May cause cancer.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Explosive
mass explosion hazard.
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 231-846-0
RTECSNumber GB2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.