Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) titanium oxide, 99.9% (metals basis)

Catalog No. p-7050122
Click to view available options
Quantity:
50 g
250 g

Catalog No. AA3567118

Supplier: Thermo Scientific Chemicals 03567118

Only null left
Add to Cart

CAS: 12060-00-3 | O3PbTi | 303.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-00-3
Molecular Formula O3PbTi
Molecular Weight (g/mol) 303.064
MDL Number MFCD00011159
InChI Key NKZSPGSOXYXWQA-UHFFFAOYSA-N
Synonym lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder,
PubChem CID 16211560
IUPAC Name dioxido(oxo)titanium;lead(2+)
SMILES [O-][Ti](=O)[O-].[Pb+2]

Specifications

Color Yellow
Physical Form Powder
Quantity 50 g
UN Number UN3288
Formula Weight 303.09
Percent Purity 99.9%
Odor Odorless
Density 7.520 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) titanium oxide
Hazard Category H300-H332-H350-H360Df-H373
Hazard Statement GHS H Statement
H300-H360-H373-H332
Fatal if swallowed.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 235-038-9
RTECSNumber XR1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.