Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Nitrate, Purified, Reagents

Catalog No. 01337196
Change view
Click to view available options
:
Quantity: 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-337-196 Quantity: 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-196 Supplier Reagents Holdings Llc Supplier No. C2229000100C2
Only null left
Add to Cart
Add to Cart

Lead Nitrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym Plumbous Nitrate, Lead(II) Nitrate, Lead Dinitrate
IUPAC Name λ²-lead(2+) dinitrate
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 4.53
Chemical Name or Material Lead Nitrate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.