Learn More
Walter Products™ Leaf Structure Model
SCI_ED
Catalog No. s07168 Shop All Walter Products Products
Click to view available options
:
Lima bean leaf model; Dimensions: 18 x 9 x 6 in. (45 x 23 x 15cm)
Vividly shows transverse and longitudinal sections along with upper and lower epidermis.
  • Lima bean leaf model
  • Mesophyll includes palisada parenchyma and spongy parenchyma
  • Surface displays stomas and guard cells
  • Details of vascular bundles, such as xylem, phloem and veins are shown
  • Enlarged 45 times life size
  • Dimensions: 18 x 9 x 6 in. (45 x 23 x 15cm)
Specifications
Material Plastic
Size 45x
Product Type Model, Leaf Structure
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.