Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEF1 Antibody (REMB6), Novus Biologicals™
SDP

Catalog No. NBP266597 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266597 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266597 Supplier Novus Biologicals Supplier No. NBP266597
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LEF1 Monoclonal specifically detects LEF1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen LEF1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone REMB6
Conjugate Unconjugated
Dilution Western Blot 5-10 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 ug/mL, Immunohistochemistry-Paraffin 1 ug/ml, Immunohistochemistry-Frozen
Formulation PBS with 0.08% Sodium Azide
Gene Accession No. P24821
Gene Alias DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF1-alpha, TCF1ALPHA
Gene Symbols LEF1
Host Species Mouse
Immunogen The immunogen for the anti-LEF1 antibody was a full length protein (Uniprot: Q9UJU2).
Purification Method Protein A or G purified
Quantity 100 μg
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing, Signal Transduction, Transcription Factors and Regulators, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 51176
Target Species Human, Mouse
Content And Storage Store at 4 °C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.