Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEF1 Mouse anti-Human, Clone: rLEF1/6906, Novus Biologicals™
SDP

Catalog No. NB177944 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177944 100 μg
NB177945 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177944 Supplier Novus Biologicals Supplier No. NBP313994100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LEF1 Monoclonal antibody specifically detects LEF1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen LEF1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rLEF1/6906
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. Q9UJU2
Gene Alias DKFZp586H0919, FLJ46390, LEF-1, lymphoid enhancer-binding factor 1, T cell-specific transcription factor 1-alpha, TCF1-alpha, TCF1ALPHA
Host Species Mouse
Immunogen Recombinant fragment (around aa100-200) of human LEF1 protein (exact sequence is proprietary) (Uniprot: Q9UJU2)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA Repair, DNA Replication Transcription Translation and Splicing, Signal Transduction, Transcription Factors and Regulators, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 51176
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.