Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lefty-1 Rat anti-Mouse, DyLight 650, Clone: RM0121-8B65, Novus Biologicals™
SDP

Catalog No. NBP122499C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122499C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122499C Supplier Novus Biologicals Supplier No. NBP122499C

Rat Monoclonal Antibody

Lefty-1 Monoclonal antibody specifically detects Lefty-1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Lefty-1
Applications Western Blot
Classification Monoclonal
Clone RM0121-8B65
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AI450052, EBAF, Leftb, lefty, lefty-1, Stra3, TGFB4
Gene Symbols LEFTY1
Host Species Rat
Immunogen Mouse recombinant protein of Lefty-1
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10637
Test Specificity This was produced from a hybridoma (mouse myeloma0 fused with spleen cells from a rat immunized with mouse recombinant protein of Lefty-1.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.