Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lefty-A Antibody (10G2), Novus Biologicals™
SDP

Catalog No. NB298637 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298637 100 μL
NB298636 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298637 Supplier Novus Biologicals Supplier No. NBP326170100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Lefty-A Monoclonal antibody specifically detects Lefty-A in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Lefty-A
Applications Western Blot, ELISA
Classification Monoclonal
Clone 10G2
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EBAFTGF-beta-4, endometrial bleeding associated factor, Endometrial bleeding-associated factor, LEFTAProtein lefty-A, left-right determination factor 2, Left-right determination factor A, LEFTYATransforming growth factor beta-4, MGC46222, Protein lefty-2, TGFB4endometrial bleeding associated factor (left-right determination, factor A,transforming growth factor beta superfamily)
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Lefty-A [UniProt O00292]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7044
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.