Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Legionella Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017622 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017622 0.1 mL
NB10064629 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017622 Supplier Novus Biologicals Supplier No. NB100646290.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Legionella Polyclonal specifically detects Legionella in Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen Legionella
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 4.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:400-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:200-1:800
Formulation PBS (pH 7.2) with 0.09% Sodium Azide
Host Species Rabbit
Immunogen A whole cell preparation of Legionella pneumophilia; ATCC#33152
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes Legionella species. This is not absorbed, so may cross-react with related microorganisms.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.