Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Legionella Antibody (LP46) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10064479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064479 0.1 mg
NB017585 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064479 Supplier Novus Biologicals Supplier No. NB10064479
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Legionella Monoclonal specifically detects Legionella in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Legionella
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone LP46
Concentration 2.8 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000
Host Species Mouse
Immunogen Native
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Strongly reactive with SG1 (Pontiac 1, Benidorm, Knoxville). Also reacts with SG1 (Philadelphia). No other serogroups reactive. No reactivity with L. erythra, L. dumoffii, E. coli or Pseudomonas aeruginosa.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.