Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Legionella Pneumophila Antibody (2F10), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB359894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359894 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359894 Supplier Novus Biologicals Supplier No. NB1102520AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Legionella Pneumophila Monoclonal antibody specifically detects Legionella Pneumophila in Bacteria samples. It is validated for ELISA

Specifications

Antigen Legionella Pneumophila
Applications ELISA
Classification Monoclonal
Clone 2F10
Conjugate Alexa Fluor 532
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias L. pneumophila, Legionellosis
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with membrane fraction of Legionella pneumophila, strain Philadelphia 1.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.