Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Legionella Pneumophila Antibody (LP101), Alexa Fluor™ 594
SDP

Catalog No. N1064476A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1064476A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1064476A94 Supplier Novus Biologicals Supplier No. NB10064476AF594

Mouse Monoclonal Antibody

Legionella Pneumophila Monoclonal specifically detects Legionella Pneumophila in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Legionella Pneumophila
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone LP101
Conjugate Alexa Fluor 594
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Legionella pneumophila serogroups 4,10,14.
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity NB100-64476 is specific for Legionalla pneumophila SS4 (Los Angeles 1). This antibody also has some minor reactivity with SG14 (1586-SCT-H) and SG10 (Leiden). There is no cross reactivity with SG1, 3, 5, 6, 7, 9, 11, 12 or 13, Listeria erythra, Listeria dumoffii, Escherichia coli or Pseudomonas aeruginosa.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.