Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Legumain/Asparaginyl Endopeptidase Antibody (46I3), PE, Novus Biologicals™
SDP

Catalog No. NB360833 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360833 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360833 Supplier Novus Biologicals Supplier No. NBP221996PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Legumain/Asparaginyl Endopeptidase Monoclonal antibody specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Legumain/Asparaginyl Endopeptidase
Applications Western Blot, ELISA
Classification Monoclonal
Clone 46I3
Conjugate PE
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34
Host Species Mouse
Immunogen Human recombinant Legumain/Asparaginyl Endopeptidase
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5641
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.