Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LELP1 Antibody (OTI3D5), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP272412C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272412C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272412C Supplier Novus Biologicals Supplier No. NBP272412C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LELP1 Monoclonal specifically detects LELP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen LELP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3D5
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias late cornified envelope-like proline-rich 1, novel small proline-rich protein
Gene Symbols LELP1
Host Species Mouse
Immunogen Full length recombinant protein of human LELP1 (NP_001010857) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 149018.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.