Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEM3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB343974 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343974 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343974 Supplier Novus Biologicals Supplier No. NBP241144MFV500

Chicken Polyclonal Antibody

LEM3 Polyclonal antibody specifically detects LEM3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen LEM3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias ANKRD41, ankyrin repeat and LEM domain containing 1, ankyrin repeat and LEM domain-containing protein 1, FLJ39369, LEM domain containing 6, LEMD6, LEM-domain containing protein 3
Host Species Chicken
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ANKLE1. The immunogen is located within the last 50 amino acids of ANKLE1. Amino Acid Squence: CAGWPPARRRRLGVHL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 126549
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.