Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEMD2 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB185311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185311 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185311 Supplier Novus Biologicals Supplier No. NBP282103H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LEMD2 Polyclonal antibody specifically detects LEMD2 in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen LEMD2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS without Preservative
Gene Alias LEM domain containing 2
Host Species Rabbit
Immunogen LEMD2 antibody was raised against a 16 amino acid synthetic peptide near the center of human LEMD2. The immunogen is located within amino acids 220 - 270 of LEMD2. Amino Acid Squence: MGKPSAPQEAEDNMKL
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221496
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.