Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Lenz Law Demonstration Apparatus
Shop All Eisco Products

Click to view available options
Class Size:
1 Lab Group
Description
- Teaches Faraday law of induction and Lenz law
- Constructed from cast iron base and aluminum ring
- Shows how passing magnet through complete loop causes device move
- No movement occurs when using the split loop
Specifications
Specifications
Grade | 9 to 12 |
For Use With (Application) | Teaches Faraday law of induction and Lenz law |
Includes | 1 Lenz' Law Apparatus |
Material | Cast Iron (Base), Aluminum (Ring) |
Class Size | 1 Lab Group |
Product Type | Lenz Law Demonstration Apparatus (Open and Closed Loop) |
Description | Demonstrates the induction of an magnetic field |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction