Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEPREL1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB134195 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB134195 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB134195 Supplier Novus Biologicals Supplier No. NBP306245AF488

Rabbit Polyclonal Antibody

LEPREL1 Polyclonal specifically detects LEPREL1 in Human samples. It is validated for Western Blot.

Specifications

Antigen LEPREL1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ10718, leprecan-like 1, leprecan-like protein 1, MCVD, myxoid liposarcoma associated protein 4, myxoid liposarcoma-associated protein 4, P3H2, prolyl 3-hydroxylase 2, prolyl 3-hydroxylase 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human LEPREL1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55214
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.