Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leptin/OB Antibody (11), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP105047AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105047AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105047AH Supplier Novus Biologicals Supplier No. NBP105047AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for ELISA.

Specifications

Antigen Leptin/OB
Applications ELISA
Classification Monoclonal
Clone 11
Conjugate Alexa Fluor 647
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS
Gene Symbols LEP
Host Species Mouse
Immunogen Recombinant human leptin adsorbed onto aluminum hydroxide gel
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3952
Test Specificity Clone 11 binds human leptin Cross-reactivity with other species has not been determined
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.