Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leptin/OB Antibody (6), Biotin, Novus Biologicals™
SDP

Catalog No. NBP221619B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221619B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221619B Supplier Novus Biologicals Supplier No. NBP221619B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Leptin/OB
Applications Western Blot, ELISA
Classification Monoclonal
Clone 6
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS
Gene Symbols LEP
Host Species Mouse
Immunogen Recombinant human leptin adsorbed onto aluminum hydroxide gel.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3952
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.