Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leptin/OB Antibody, Novus Biologicals™
SDP

Catalog No. NB300611 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300611 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300611 Supplier Novus Biologicals Supplier No. NB300611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Leptin/OB Polyclonal specifically detects Leptin/OB in Human, Mouse, Rat, Porcine, Avian, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry.

Specifications

Antigen Leptin/OB
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Frozen 1:50 - 1:200, Immunocytochemistry 1:50 - 1:200
Formulation PBS and 1 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. P41160
Gene Alias FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS
Gene Symbols LEP
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues Q(25) K V Q D D T K T L I K T I V T R I N D(44) of mouse Leptin.
Purification Method Affinity Purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3952
Test Specificity Leptin
Target Species Human, Mouse, Rat, Pig, Avian, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.