Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leptin R Antibody, Novus Biologicals™
SDP

Catalog No. NB10040796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10040796 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10040796 Supplier Novus Biologicals Supplier No. NB10040796
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Leptin R Polyclonal specifically detects Leptin R in Human, Mouse, Rat samples. It is validated for Control, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen Leptin R
Applications Control, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2ug/ml, Flow Cytometry 10ug/ml, Immunohistochemistry 4-6ug/ml, Immunohistochemistry-Paraffin 4-6ug/ml, Peptide ELISA Detection limit 1:128000, Control
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_002294.2
Gene Alias CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295
Gene Symbols LEPR
Host Species Goat
Immunogen Peptide with sequence C-TQDDIEKHQSDAG corresponding to internal region according to NP_002294.2, NP_001003679.1, NP_001003680.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3953
Test Specificity This antibody is expected to recognize all three reported isoforms (NP_002294.2; NP_001003679.1; NP_001003680.1).
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.