Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leptin R Antibody (RM0085-9C29), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP121725IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121725IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121725IR Supplier Novus Biologicals Supplier No. NBP121725IR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Leptin R Monoclonal specifically detects Leptin R in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Leptin R
Applications Western Blot
Classification Monoclonal
Clone RM0085-9C29
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295
Gene Symbols LEPR
Host Species Rat
Immunogen Mouse recombinant EC-domain of LeptinR
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3953
Test Specificity This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of Extracellular domain of Leptin R.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.