Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LETM2 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB043479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB043479 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB043479 Supplier Novus Biologicals Supplier No. NBP297437C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LETM2 Polyclonal specifically detects LETM2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen LETM2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FLJ25409, LETM1 and EF-hand domain-containing protein 2, LETM1 domain-containing protein LETM2, mitochondrial, leucine zipper-EF-hand containing transmembrane protein 1-like protein, leucine zipper-EF-hand containing transmembrane protein 2, Leucine zipper-EF-hand-containing transmembrane protein 1-like
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human LETM2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 137994
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.