Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leucine, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
61-90-5
C6H13NO2
131.18
ROHFNLRQFUQHCH-UHFFFAOYNA-N
2-amino-4-methylpentanoic acid
CC(C)CC(N)C(O)=O
61-90-5
C6H13NO2
131.18
ROHFNLRQFUQHCH-UHFFFAOYNA-N
2-amino-4-methylpentanoic acid
CC(C)CC(N)C(O)=O

L1202, 61-90-5, C6H13NO2

Spectrum™ Leucine, USP is a hydrophobic essential amino acid that is a component in astacin, ferritin and other buffer proteins. It is used for the creation of sterols in the muscle and adipose tissues. Widely used in sport supplements for its muscle building properties, it is also used as a food additive and flavor enhancer. Spectrum Chemical manufactured USP LEUCINE products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Chloride 0.0005
Ignition Residue 0.004
Loss on Drying 0.002
Packaging Poly Bottle
Specific Rotation +14.9° to +17.3°
Percent Purity 98.5 to 101.5%
Sulfate 0.0003
Quantity 1 kg